Imprimer

Anti-Smt3 Mouse Monoclonal Antibody [clone: N/A]

Fournisseur: Cayman Chemical
12021-500
CAYM12021-500EA 540 EUR
CAYM12021-500
Anti-Smt3 Mouse Monoclonal Antibody [clone: N/A]
Anticorps
Type: Primary
Antigen: Smt3
Clonality: Monoclonal
Clone: N/A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

SPÉCIFICATIONS

Symbole de l'antigène Smt3
Environmentally Preferable
Type d'anticorps Primaire
Clonalité Monoclonal
Conjugaison Unconjugated
Réactivité
Hôte Mouse
Isotype IgG1
Western blot Yes
ELISA Yes
Immunogène recombinant yeast Smt3 amino acids 1-97
Nombre d'acides aminés 1 to 97
Purification purified by Protein G chromatography
Adsorption croisée No
Température de stockage –20 °C
Température de transport –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR