Imprimer

Anti-PGT Rabbit Polyclonal Antibody

Fournisseur: Cayman Chemical
11860-1
CAYM11860-1EA 540 EUR
CAYM11860-1
Anti-PGT Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PGT
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat, Sheep
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR