Imprimer

Anti-Trx1 Rabbit Polyclonal Antibody

Fournisseur: Cayman Chemical
11540-1
CAYM11540-1EA 523 EUR
CAYM11540-1
Anti-Trx1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Trx1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR