Imprimer

10-Acetyl-3,7-dihydroxyphenoxazine ≥98%

Fournisseur: Cayman Chemical
Attention

Synonyms: 1-(3,7-Dihydroxy-10H-phenoxazin-10-yl)ethanone, ADHP, Amplex® Red (Registered trademark of LifeTechnologies)

10010469-10 10010469-5 10010469-1 10010469-25
CAYM10010469-10EA 224 EUR
CAYM10010469-10 CAYM10010469-5 CAYM10010469-1 CAYM10010469-25
10-Acetyl-3,7-dihydroxyphenoxazine ≥98%
10-Acetyl-3,7-dihydroxyphenoxazine

10-Acetyl-3,7-dihydroxyphenoxazine (ADHP) is a highly sensitive, stable substrate for horseradish peroxidase (HRP) that enables selective detection of H₂O₂.


This non-fluorescent reagent reacts with H₂O₂ to produce the fluorescent compound resorufin, which can be analysed using an excitation wavelength of 520 to 550 nm and an emission wavelength of 585 to 595 nm. In a 96 well plate format, ADHP enables detection of H₂O₂ at a concentration as low as 5 pmol per 100 µl sample.

Formule: C₁₄H₁₁NO₄
MW: 257.24 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00467861
Numéro CAS: 119171-73-2

Commander maintenant

Résultats des tests de spécification

Purity ≥98%
λmax 250, 285 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR