Imprimer

Anti-TXA2-R Rabbit Polyclonal Antibody

Fournisseur: Cayman Chemical
10004452-1
CAYM10004452-1EA 540 EUR
CAYM10004452-1
Anti-TXA2-R Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TXA2-R
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: cos-7, Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR