Imprimer
Flacons, aluminium, agréés UN, avec bouchon à vis
Fournisseur: BURKLE
0329-0125 0329-1250 0329-0500 0329-0250
BURK0329-0125EA 35.1 EUR
BURK0329-0125 BURK0329-1250 BURK0329-0500 BURK0329-0250
Flacons, aluminium, agréés UN, avec bouchon à vis
Flacons Flacons en métal
Pure aluminium (material AL 99,5).
  • UN code is printed onto the bottles in black
  • With eyelets, sealable (except the 125 ml model)
  • Light, unbreakable, seamless
Aluminium transport bottles with screw cap close safely and tightly and are approved for packaging type I (X).
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR