Imprimer
Evaporating flasks
Fournisseur: BUCHI
008750 008751 008754 008758 008736 008737 008738 008739 008753 008761 008762 008764 008765 008767 008770 020730 025261 025262 025263 025324 025517 027346 047990 047992
BUCH008750EA 118 EUR
BUCH008750 BUCH008751 BUCH008754 BUCH008758 BUCH008736 BUCH008737 BUCH008738 BUCH008739 BUCH008753 BUCH008761 BUCH008762 BUCH008764 BUCH008765 BUCH008767 BUCH008770 BUCH020730 BUCH025261 BUCH025262 BUCH025263 BUCH025324 BUCH025517 BUCH027346 BUCH047990 BUCH047992
Evaporating flasks
Ballons Flacons pour évaporateurs rotatifs
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® rotary evaporators. Centre neck. Pear-shaped with ground joint socket or EL-flange.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR