Imprimer
Anti-Peripherin Chicken Polyclonal Antibody
Fournisseur: Biosensis
C-1399-50
BSENC-1399-50EA 312 EUR
BSENC-1399-50
Anti-Peripherin Chicken Polyclonal Antibody
Anticorps
Peripherin is a class-III neuronal intermediate filament protein found in certain classes of neuron, most of which are located in the peripheral nervous system. Peripherin is a ~57kDa intermediate filament subunit found initially in sensory neurons of the peripheral nervous systems, which gives the protein its name. The HGNC name for this protein is PRPH. Subsequently, peripherin was found in some sensory and other neurons of the central nervous system and also in rat pheochromacytoma PC12 cells. Peripherin is also expressed in certain neuroendocrine tumors and in the insulin producing cellsMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR