Imprimer

Anti-INA Rabbit Polyclonal Antibody

Fournisseur: Boster Bio
PB10003
BSBTPB10003EA 552 EUR
BSBTPB10003
Anti-INA Rabbit Polyclonal Antibody
Anticorps
Alpha-Internexin (INA; also NF-66) is a 66 kDa member of the intermediate filament (IF) protein family. The protein was originally purified from rat optic nerve and spinal cord. And the protein copurifies with other neurofilament subunits, as it was originally discovered, however in some mature neurons it can be the only neurofilament expressed. The protein is present in developing neuroblasts and in the Central Nervous System of adults. Meanwhile, the protein is a major component of the intermediate filament network in small interneurons and cerebellar granule cells, where it is present inMore Product Information
Commander maintenant

SPÉCIFICATIONS

Symbole de l'antigène INA
Environmentally Preferable
Nom de l'antigène internexin neuronal intermediate filament protein, alpha
Synonymes antigène NEF5|TXBP-1|NF-66
Type d'anticorps Primaire
Clonalité Polyclonal
Conjugaison Unconjugated
Réactivité Human,Rat,Mouse
Hôte Rabbit
ID de gène 9118
Isotype IgG
Western blot Yes
ImmunoChimie Yes
Immunogène E.coli-derived human Alpha Internexin recombinant protein (Position: A71-R161). Human Alpha Internexin shares 98.9% amino acid (aa) sequence identity with both mouse and rat Alpha Internexin.
Formulaire Lyophilized
Adsorption croisée No
Température de stockage At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR