Imprimer
MALDI AnchorChip™ Targets
Fournisseur: BRUKER DALTRONIK
8280790
BRUK8280790EA 3640 EUR
BRUK8280790
MALDI AnchorChip™ Targets
Microplaques Microplaques immunologiques et bandelettes
Patented AnchorChip™ targets are polished steel targets with hydrophilic sample and calibrant spots surrounded by a hydrophobic barrier for MALDI sample preparation.
  • Spot Ø: 800 µm
  • Sample spots: 384
  • Calibrant spots: 96
  • Autoteaching
Sample positions on contain 'anchors', hydrophilic patches surrounded by a hydrophobic ring. The 'anchor' localises droplets at the sample position and the hydrophobic ring prevents sample spreading and concentrates the sample into a spot between 0,4 and 2 mm in diameter. The localisation effect ensures that every singleMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR