Imprimer
Absorbent pillows, ALLWICK Universal
Fournisseur: Brady
813792
BRDY813792EA 181.48 EUR
BRDY813792
Absorbent pillows, ALLWICK Universal
Matériaux absorbants
These absorbent pillows absorb spills and leaking fluids in tight spaces and under persistent drip sources.
  • Absorbs a variety of liquids including water, coolants, solvents, oils, antifreeze, etc
  • Ideal under machines or in tight spaces
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR