Imprimer
Thermal transfer printer ribbon, R6606 series
Fournisseur: Brady
622116
BRDY622116EA 78.59 EUR
BRDY622116
Thermal transfer printer ribbon, R6606 series
Rubans pour imprimante
Diesel-resistant thermal transfer ribbon on Brady PermaSleeve ™ and Heatex ™ heat shrink tubing. This print ribbon is highly resistant to chemicals and friction.
Conditionnement: One print ribbon/box.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR