Imprimer
Aggressive Adhesive Multi-Purpose Nylon Labels B-499 for M410, BMP41, M510, BMP51, BMP53, M511 Printers
Fournisseur: Brady
170799 170801 170931 170932 170933 170927 170807 170808 170793 170794 170795 170796 170797 170798 170800 170802 170803 170804 170805 170806 170930 170934
BRDY170799EA 53.73 EUR
BRDY170799 BRDY170801 BRDY170931 BRDY170932 BRDY170933 BRDY170927 BRDY170807 BRDY170808 BRDY170793 BRDY170794 BRDY170795 BRDY170796 BRDY170797 BRDY170798 BRDY170800 BRDY170802 BRDY170803 BRDY170804 BRDY170805 BRDY170806 BRDY170930 BRDY170934
Aggressive Adhesive Multi-Purpose Nylon Labels B-499 for M410, BMP41, M510, BMP51, BMP53, M511 Printers
Étiquettes
For a tough and versatile general-purpose label material, look no further than the B-499 nylon cloth label material. Print almost anywhere with a Brady portable label printer.
  • General-purpose nylon cloth material
  • For smooth, textured/rough, or highly textured/low surface energy surfaces
  • Designed to withstand extreme temperatures, chemicals, greases, oils, cleaners and detergents
  • Permanent adhesive labels stick to smooth, rough, and powder-coated surfaces
This versatile selection of Aggressive Adhesive Multi-Purpose Nylon Labels are ideal for labellingMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR