Imprimer
Gauze, Askina®
Fournisseur: B. Braun
9030409W
BRAU9030409WEA 166 EUR
BRAU9030409W
Gauze, Askina®
Matériel de premiers secours Pansements
Absorbent gauze for making compresses and swabs, as an ointment or medication carrier.
  • 80 cm wide, folded into 8 layers to a width of 10 cm
  • Bleached 100% without chlorine
  • Highly absorbent
  • Can be cut to size
Certifications: DIN 61630 - VM 20
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR