Imprimer
Cell disruptor with handheld converter and ¹/₈" microtip, Sonifier® SFX150
Fournisseur: BRANSON
101-063-1097R
BRAN101-063-1097REA 4920 EUR
BRAN101-063-1097R
Cell disruptor with handheld converter and ¹/₈" microtip, Sonifier® SFX150
Homogénéisateurs Désintégrateurs de cellules à ultrasons
The SFX150 Branson's handheld converter is designed with the user in mind with an ergonomic grip for comfort and control, and a recessed pushbutton and LED indicator for assured operation and to bring the industry's most advanced sample-processing capabilities to your laboratory.
  • Automatically adjust the ultrasonic pulse length
  • Prevent the sample from overheating (requires optional temperature probe)
  • Microtip limit 70%
  • Temperature control allows the user to specify a temperature range for the sample
Applications include sample preparation, cellMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR