Imprimer

Anti-MAPK9 Rabbit Polyclonal Antibody (Cy5®)

Fournisseur: Bioss
BS-5487R-CY5
BOSSBS-5487R-CY5EA 930 EUR
BOSSBS-5487R-CY5
Anti-MAPK9 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
phosphorylated at the Thr-Pro-Tyr phosphorylation motif instead of the characteristic MAP kinase Thr-Glu-Tyr motif. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAPK9
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR