Imprimer
Anti-GPR128 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-13518R-A750
BOSSBS-13518R-A750EA 890 EUR
BOSSBS-13518R-A750
Anti-GPR128 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
G protein-coupled receptors (GPCRs), also designated seven transmembrane (7TM) receptors and heptahelical receptors, are a protein family which interact with G proteins (heterotrimeric GTPases) to synthesize intracellular second messengers such as diacylglycerol, cyclic AMP, inositol phosphates, and calcium ions. Their diverse biological functions range from vision and olfaction to neuronal and endocrine signaling and are involved in many pathological conditions. G protein receptor 128 (GPR128), a member of the secretin family of GCPRs with a GPS domain in its N-terminal domain, may mediateMore Product Information
Plus About VWR