Imprimer
Anti-GPR84 Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-13507R-CY5
BOSSBS-13507R-CY5EA 900 EUR
BOSSBS-13507R-CY5
Anti-GPR84 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
G protein-coupled receptors (GPCRs), also designated seven transmembrane (7TM) receptors and heptahelical receptors, are a protein family which interact with G proteins (heterotrimeric GTPases) to synthesize intracellular second messengers such as diacylglycerol, cyclic AMP, inositol phosphates, and calcium ions. Their diverse biological functions range from vision and olfaction to neuronal and endocrine signaling and are involved in many pathological conditions. G protein receptor 84 (GPR84), a member of the GCPR 1 family, is an orphan GCPR expressed in bone marrow, brain, heart, muscle,More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR