Imprimer
Anti-MEGF10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-12372R-A750
BOSSBS-12372R-A750EA 890 EUR
BOSSBS-12372R-A750
Anti-MEGF10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
MEGF10 is a 1140 amino acid protein encoded by the human gene MEGF10. MEGF10 belongs to the MEGF family and contains fifteen EGF-like domains and one EMI domain. The engulfment of dying cells is a specialised form of phagocytosis that is highly conserved across evolution and is mediated by the concerted action of several molecules. MEGF10 is an engulfment receptor protein that localizes to the plasma membrane in a punctuated pattern. The MEGF10 protein shares structural similarities with nematode engulfment receptor cell death abnormal-1 (CED-1). MEGF10 function is modulated by the ATPMore Product Information
Plus About VWR