Imprimer

Anti-5-HT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: Bioss
BS-1126R-HRP
BOSSBS-1126R-HRPEA 883 EUR
BOSSBS-1126R-HRP
Anti-5-HT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Serotonin (pronounced) (5-hydroxytryptamine, or 5-HT) is a monoamine neurotransmitter synthesized in serotonergic neurons in the central nervous system (CNS) and enterochromaffin cells in the gastrointestinal tract of animals including humans. Serotonin is also found in many mushrooms and plants, including fruits and vegetables.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: 5-HT
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR