Imprimer
Filtering tiles, BOLA
Fournisseur: Bohlender
N1610-10 N1616-30 N1616-20 N1616-10
BOHLN1610-10EA 754 EUR
BOHLN1610-10 BOHLN1616-30 BOHLN1616-20 BOHLN1616-10
Filtering tiles, BOLA
Filtres Carrés filtrants
These filtering tiles are made of microporous PTFE.
  • Suitable for cutting or stamping
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR