Imprimer

Screw caps with aperture, BOLA

Fournisseur: Bohlender
H999-45
BOHLH999-45EA 3.8 EUR
BOHLH999-45
Screw caps with aperture, BOLA
Capuchons Bouchons pour flacons
Screw caps, made of PP, with handy knurl.

  • Diameter of aperture: 34 mm
  • Good chemical resistance
  • Temperature resistance: from –20 to +110 °C
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR