Imprimer
Vacuum stopcocks, BOLA
Fournisseur: Bohlender
B293-02
BOHLB293-02EA 488 EUR
BOHLB293-02
Vacuum stopcocks, BOLA
Robinets d'arrêt
Made of PTFE.
  • Universal chemical resistance
  • Temperature resistance: from 0 to +110 °C
Vacuum stopcock with ground joint cone, bore and connections for tubing. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR