Imprimer
Double bosshead, cross type
Fournisseur: BOCHEM
5333
BOCH5333EA 17.1 EUR
BOCH5333
Double bosshead, cross type
Pinces Noix de serrage
Cross type bossheads made of aluminium.
  • For rods up to 16,5 mm
  • Angle: 90°
  • With thumb screw (M8)
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR