Imprimer
Forceps, titanium
Fournisseur: BOCHEM
1910
BOCH1910EA 69 EUR
BOCH1910
Forceps, titanium
Pinces
These titanium forceps are particularly suitable for applications where high corrosion-resistance, strength and low weight are important.
  • Lightweight, high strength
  • Corrosion and thermal resistance
  • Resistant to diluted sulphuric acid, hydrochloric acid, solutions containing chloride, cold nitric acid as well as most organic acids and alkaline solutions, such as sodium hydroxide
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR