Imprimer
Transport trolleys for baskets, EURO-STANDARD
Fournisseur: BOCHEM
17333
BOCH17333EA 1290 EUR
BOCH17333
Transport trolleys for baskets, EURO-STANDARD
Chariots
Transport cart made of stainless steel used for transport baskets.
  • Five hanging ties
  • Two pushing handles
  • Two guide and fixed rolls, two with lock
  • Enclosure of rolls, zincked
  • Excess length of enclosure
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR