Imprimer
Hand dynamometer, Smedley spring-type, Baseline®
Fournisseur: 3B Scientific
W54654
BINHW54654EA 0 EUR
BINHW54654
Hand dynamometer, Smedley spring-type, Baseline®
Dynamomètres
Digital, lightweight Smedley spring dynamometer offers accurate grip strength readout.
  • 5-preset grip sizes
  • Captures and displays maximum grip force in both kilogrammes and pounds
  • Unit assesses results according to age and gender
  • Unit saves/stores results for fast retrieval with up to 19 users
  • The adjustable handle comfortably accommodates any hand size.
Information de livraison: Supplied with protective carrying case.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR