Imprimer
Microscope slides with printed rings
Fournisseur: BELLCO GLASS
5638-01920
BELC5638-01920EA 164.51 EUR
BELC5638-01920
Microscope slides with printed rings
Lames Lames de microscopes
These microscopy slides with three printed rings allow for counting bacteria by spreading a known volume of solution over a known area for the calculation of bacteria per sample.
Area (ring): 1 cm²
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR