Imprimer
Battery powered engraving tool, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F441500005
BELAF441500005EA 64.9 EUR
BELAF441500005
Battery powered engraving tool, SP Bel-Art
Stylos Stylets et traceurs
Battery powered engraving tool to permanently mark most materials including glass, plastic, wood, metal, bone, and stone.
  • Cordless battery power makes it portable for use virtually anywhere indoors or out in the field
  • Marks will not fade if exposed to chemicals or sunlight
  • Operates on two 'AA' batteries
Simply hold the engraving tool like a pen, press the power button, apply light pressure and write on test tubes, bottles, racks, keys, equipment, tools, electronics and more.
Informations de commande: Convenient polyethylene stand (sold separately)More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR