Imprimer
Supports for round bottom flasks, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F389500000
BELAF389500000EA 104 EUR
BELAF389500000
Supports for round bottom flasks, SP Bel-Art
Supports Supports pour ballons
This PP round bottom flask support fits any size round-bottom flask up to 10 liters using stepped, concentric rings.
  • Stackable
  • Hands-free, upright position
  • Good chemical and temperature resistance
Use several for adjustable work height.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR