Imprimer
Spill containment tray with grid, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F246760003
BELAF246760003EA 221 EUR
BELAF246760003
Spill containment tray with grid, SP Bel-Art
Plateaux/Récipients/Pots Plateaux de déversements
This large, deep, polypropylene plastic tray protects users by capturing accidental hazardous spills.
  • High chemical resistance
  • Epoxy coated wire support grid holds numerous vessels or racks of test tubes
  • Steam autoclavable at 121 °C
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR