Imprimer
Drying tubes with tube fittings, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F199600000 F199610000 F199620000
BELAF199600000EA 60.5 EUR
BELAF199600000 BELAF199610000 BELAF199620000
Drying tubes with tube fittings, SP Bel-Art
Tubes Tubes de dessication
Unbreakable polyethylene straight tubes.
  • Good chemical resistance
  • Godd temperature resistance
  • Drying agent no included
Can be filled with desiccant of your choice. Each tube is supplied with two polypropylene fittings which accept 6,4 to 9,5 mm int.Ø tubing.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR