Imprimer
Microcentrifuge tube opener, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F199250001
BELAF199250001EA 61.4 EUR
BELAF199250001
Microcentrifuge tube opener, SP Bel-Art
Pinces à sertir et pinces à dessertir
Easily open all sizes of microcentrifuge tubes with this double-ended tool.
  • Opens standard 1,5, 2,0 and 2,2 ml tubes with one end
  • Forked blade on other end opens smaller sizes (250, 400 and 550 μl)
  • Dimensions: 7 cm (3⅛") long
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR