Imprimer
Hybridisation bottle racks, Poxygrid®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F169640006
BELAF169640006EA 250 EUR
BELAF169640006
Hybridisation bottle racks, Poxygrid®, SP Bel-Art
Portoirs Portoirs pour flacons
Racks made of epoxy-coated welded steel.
  • Easy cleaning
  • Provides maximum protection
  • Good chemical and temperature resistance
It holds hybridisation bottles of 40 mm diameter and 150/300 mm long bottles.
W×D×H: 137×198×130 mm
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR