Imprimer
Soxhlet extractor, multiposition
Fournisseur: BEHR
B00218423
BEHRB00218423EA 8380 EUR
BEHRB00218423
Soxhlet extractor, multiposition
Extracteurs Extracteurs Soxhlet
Complete workstation to undertake Soxhlet extraction using 6×100 ml flasks.
Information de livraison: Workstation comprises temperature and time controller, heating block with six sample positions, condenser frame with coolant water distribution manifold, glassware including flasks, extractors and Dimroth condensers.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR