Imprimer
Neutralisation unit, SIMVA, behrotest®
Fournisseur: BEHR
B00217922
BEHRB00217922EA 644 EUR
BEHRB00217922
Neutralisation unit, SIMVA, behrotest®
Extracteurs de vapeur
For behr digestion units (K-block & Inkjel devices), extraction systems for acid vapours are a necessity. We differentiate between a simple water jet pump / SIMVAC and the behrosog3 extraction system and neutraliser.
  • SIMVAC can be used for digestion blocks with up to 8 sample positions
  • Only requires a water connection to generate the required extraction performance – nothing else
  • Easy, robust and price effective
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR