Imprimer
Flow meter
Fournisseur: BEHR
B00217617
BEHRB00217617EA 556 EUR
BEHRB00217617
Flow meter
Débitmètres
This adjustable flow through meter is a substitute of DFM for the determination of cyanide.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR