Imprimer

β-Cyclodextrine

Fournisseur: Beckman Coulter

Synonyms: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex

713331
BECL713331EA 0 EUR
BECL713331
β-Cyclodextrine
β-Cyclodextrine
Formule: C₄₂H₇₀O₃₅
MW: 1134.98 g/mol
Numéro MDL: MFCD00078139
Numéro CAS: 7585-39-9
EINECS: 231-493-2

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR