Imprimer
Experimentation kits, dot blot analysis
Fournisseur: G-Biosciences
BE-502
BE-502EA 384 EUR
BE-502
Experimentation kits, dot blot analysis
Matériel éducatif Matériel éducatif de biologie Activités de physiologie et santé
Dot blotting is a simple technique to identify a known protein in a biological sample. The ease and simplicity of the technique makes dot blotting an ideal diagnostic tool.
  • Simulated test samples
  • Teaches immunodetection technique
  • Hands-on Dot Blot Analysis technique with necessary reagents and supplies
The method involves applying small volumes of antigens or test samples on protein binding membrane; the membrane captures and immobilizes proteins. The proteins immobilized on the membrane are probed with a specific antibody and a matching specificMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR