Imprimer
BD Q-Syte™ Needle-Free Connector
Fournisseur: BD Medical
385100 385151 380510
BDAM385100EA 192 EUR
BDAM385100 BDAM385151 BDAM380510
BD Q-Syte™ Needle-Free Connector
Canules
The BD Q-Syte™ Needle-free connector has a split septum design that complies with the guidelines for reducing catheter-related bloodstream infection (CRBSI) rates set by the Centers for Disease Control and Prevention (CDC) and the International Nosocomial Infection Control Consortium (INICC).
  • Lower CRBSI rates than complex mechanical valve systems
  • Clear housing allows visual assessment of the fluid path
  • Straight, open fluid path design provides a high flow rate
Research shows that split septum needle-free systems have 64 to 70% lower CRBSI rates thanMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR