Imprimer

6-Hydroxyindole, N-BOC protected 95%

Fournisseur: Apollo Scientific

Synonyms: tert-Butyl 6-hydroxy-1H-indole-1-carboxylate

OR2572-1G
APOSOR2572-1GEA 151 EUR
APOSOR2572-1G
6-Hydroxyindole, N-BOC protected 95%
6-Hydroxyindole, N-BOC protected
Formule: C₁₃H₁₅NO₃
MW: 233.263 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD08272252
Numéro CAS: 898746-82-2

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR