Imprimer
Bravo NGS Workstation
Fournisseur: AGILENT
G5574AA
AGLSG5574AAEA 363281 EUR
AGLSG5574AA
Bravo NGS Workstation
Instruments de préparation d'échantillons
High-throughput (HT) NGS sample preparation robot for increased efficiency and maximised walk away time.
  • Quickly automate with ready-to-use protocols and pre-developed methods for Agilent SureSelect, Agilent HaloPlex, Illumina TruSeq, NuGEN Ovation, KAPA HTP, PacBio SMRTbell, NEBNext, and more
  • Easy-to-use interface requires no programming expertise to run your automated methods
  • Consistent data quality allows you to increase your throughput, with consistent results every time
  • Prepare up to ten times more samples in the same time
  • Walk-away automation reduces
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR