Imprimer

Anti-PI3KC2G Rabbit Polyclonal Antibody

Fournisseur: Abgent
AP8012A
ABGEAP8012AEA 354 EUR
ABGEAP8012A
Anti-PI3KC2G Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PI3KC2G
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR