Imprimer

Anti-MAPK10 Rabbit Polyclonal Antibody

Fournisseur: Abgent
AP7222A
ABGEAP7222AEA 354 EUR
ABGEAP7222A
Anti-MAPK10 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: MAPK10 (mitogen-activated protein kinase 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR