Imprimer

Anti-PRKCA Rabbit Polyclonal Antibody

Fournisseur: Abgent
AP7015A
ABGEAP7015AEA 354 EUR
ABGEAP7015A
Anti-PRKCA Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PRKCA (protein kinase C, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR