Imprimer

Anti-DXO Rabbit Polyclonal Antibody

Fournisseur: Abgent
AP6635A
ABGEAP6635AEA 354 EUR
ABGEAP6635A
Anti-DXO Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: DXO (decapping exoribonuclease)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR