Imprimer

Anti-TEC Rabbit Polyclonal Antibody

Fournisseur: Abgent
AP16402A
ABGEAP16402AEA 354 EUR
ABGEAP16402A
Anti-TEC Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TEC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR