Imprimer

Anti-ABCC12 Rabbit Polyclonal Antibody

Fournisseur: Abgent
AP13196C
ABGEAP13196CEA 354 EUR
ABGEAP13196C
Anti-ABCC12 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: ABCC12 (ATP-binding cassette, sub-family C (CFTR/MRP), member 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR