Imprimer

Anti-HEPACAM Rabbit Polyclonal Antibody

Fournisseur: Abgent
AP12843B
ABGEAP12843BEA 354 EUR
ABGEAP12843B
Anti-HEPACAM Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: HEPACAM (hepatic and glial cell adhesion molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR