Imprimer
Buccutite™ Rapid PE-Cy5 Tandem Antibody Labelling Kit
Fournisseur: AAT BIOQUEST
5416
AATB5416EA 2390 EUR
AATB5416
Buccutite™ Rapid PE-Cy5 Tandem Antibody Labelling Kit
Systèmes de liaison et de marquage des protéines
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ Rapid PE-Cy5 tandem antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PE, APC, PerCP and iFluor® tandem dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR